Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123829.1 | NP_001117301.1 | 10 - 11 | 1015 | 92 | |
NM_101598.3 | NP_564018.1 | 10 - 11 | 1130 | 92 | ||
GenBank mRNA | AY085705.1 | - | 10 - 11 | 932 | 92 | |
BT026477.1 | - | 10 - 11 | 812 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |