Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123829.1 | NP_001117301.1 | 8 - 9 | 815 | 65 | |
NM_101598.3 | NP_564018.1 | 8 - 9 | 921 | 65 | ||
GenBank mRNA | AY085705.1 | - | 8 - 9 | 723 | 65 | |
BT026477.1 | - | 8 - 9 | 603 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |