Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101598.3 | NP_564018.1 | 9 - 10 | 1043 | 72 | |
GenBank mRNA | AY085705.1 | - | 9 - 10 | 845 | 72 | |
BT026477.1 | - | 9 - 10 | 725 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |