Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332318.1 | NP_001321828.1 | 10 - 11 | 1088 | 79 | |
NM_101628.4 | NP_564030.2 | 10 - 11 | 973 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |