Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101639.3 | NP_173218.2 | 6 - 7 | 880 | 109 | |
GenBank mRNA | BT002320.1 | - | 6 - 7 | 534 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |