Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101651.4 | NP_173230.1 | 5 - 6 | 433 | 94 | |
GenBank mRNA | AF332407.1 | - | 4 - 5 | 316 | 94 | |
AJ242970.1 | - | 5 - 6 | 370 | 94 | ||
AY045793.1 | - | 5 - 6 | 402 | 94 | ||
AY079361.1 | - | 4 - 5 | 316 | 94 | ||
AY084522.1 | - | 5 - 6 | 404 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |