Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMPK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198098.1 | NP_001185027.1 | 7 - 8 | 1326 | 85 | |
NM_101675.4 | NP_173253.1 | 7 - 8 | 1338 | 85 | ||
NM_179354.3 | NP_849685.1 | 6 - 7 | 1418 | 85 | ||
GenBank mRNA | AY045931.1 | - | 7 - 8 | 1248 | 85 | |
AY142618.1 | - | 6 - 7 | 1170 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |