Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332357.1 | NP_001321210.1 | 5 - 6 | 1733 | 66 | |
NM_001332358.1 | NP_001321211.1 | 5 - 6 | 1552 | 66 | ||
NM_101725.3 | NP_173302.2 | 5 - 6 | 1733 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |