Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101746.5 | NP_564066.2 | 1 - 2 | 991 | 84 | |
GenBank mRNA | AK227602.1 | - | 1 - 2 | 878 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |