Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332406.1 | NP_001323012.1 | 3 - 4 | 955 | 119 | |
NM_101834.4 | NP_173408.2 | 5 - 6 | 1571 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |