Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iqd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332417.1 | NP_001320832.1 | 1 - 2 | 1278 | 83 | |
NM_101842.5 | NP_564097.1 | 2 - 3 | 969 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |