Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101847.4 | NP_564099.1 | 4 - 5 | 1463 | 102 | |
GenBank mRNA | U06276.1 | - | 4 - 5 | 1335 | 102 | |
U40715.1 | - | 4 - 5 | 1355 | 102 | ||
X79210.1 | - | 1340 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |