Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101863.4 | NP_173437.1 | 4 - 5 | 732 | 87 | |
GenBank mRNA | AF332438.1 | - | 4 - 5 | 351 | 87 | |
AF360154.1 | - | 4 - 5 | 621 | 87 | ||
AK227384.1 | - | 4 - 5 | 619 | 87 | ||
AY056308.1 | - | 4 - 5 | 351 | 87 | ||
U45236.1 | - | 4 - 5 | 610 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |