Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101884.4 | NP_173458.1 | 1 | 1250 | 104 | |
GenBank mRNA | AY046042.1 | - | 1 | 1143 | 104 | |
AY113961.1 | - | 1 | 1072 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |