Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCB2;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101912.3 | NP_173485.1 | 6 - 7 | 733 | 111 | |
GenBank mRNA | BT029740.1 | - | 5 - 6 | 426 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |