Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035995.4 | NP_001031072.3 | 2 - 3 | 1454 | 106 | |
NM_001035996.1 | NP_001031073.1 | 3 - 4 | 470 | 106 | ||
NM_001332452.1 | NP_001323332.1 | 3 - 4 | 1353 | 106 | ||
NM_001332453.1 | NP_001323333.1 | 1 - 2 | 937 | 106 | ||
NM_101913.4 | NP_564120.1 | 3 - 4 | 565 | 106 | ||
NM_202144.2 | NP_973873.1 | 3 - 4 | 470 | 106 | ||
GenBank mRNA | AK316990.1 | - | 3 - 4 | 475 | 106 | |
AY056447.1 | - | 3 - 4 | 449 | 106 | ||
AY058104.1 | - | 3 - 4 | 452 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |