Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035995.4 | NP_001031072.3 | 5 - 6 | 2480 | 67 | |
NM_001332452.1 | NP_001323332.1 | 6 - 7 | 2379 | 67 | ||
NM_001332453.1 | NP_001323333.1 | 2 - 3 | 2147 | 67 | ||
NM_101913.4 | NP_564120.1 | 6 - 7 | 1591 | 67 | ||
NM_202144.2 | NP_973873.1 | 5 - 6 | 1595 | 67 | ||
GenBank mRNA | AY056447.1 | - | 6 - 7 | 1475 | 67 | |
AY058104.1 | - | 6 - 7 | 1478 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |