Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101914.4 | NP_564121.1 | 5 - 6 | 1436 | 66 | |
GenBank mRNA | AK227182.1 | - | 5 - 6 | 1343 | 66 | |
AY136424.1 | - | 5 - 6 | 1342 | 66 | ||
BT010373.1 | - | 4 - 5 | 1260 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |