Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332471.1 | NP_001319055.1 | 5 - 6 | 1850 | 83 | |
NM_101937.6 | NP_173508.1 | 4 - 5 | 2040 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |