Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101938.5 | NP_564126.1 | 3 - 4 | 953 | 59 | |
GenBank mRNA | AF191028.1 | - | 3 - 4 | 861 | 59 | |
AK227749.1 | - | 3 - 4 | 889 | 59 | ||
BT004822.1 | - | 3 - 4 | 856 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |