Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKB2;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101946.4 | NP_173517.1 | 6 - 7 | 792 | 86 | |
GenBank mRNA | AK229456.1 | - | 6 - 7 | 791 | 86 | |
BT024780.1 | - | 6 - 7 | 710 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |