Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101951.4 | NP_173522.1 | 9 - 10 | 3257 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |