Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332488.1 | NP_001322652.1 | 3 - 4 | 661 | 149 | |
NM_101971.3 | NP_173541.1 | 3 - 4 | 881 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |