Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332504.1 | NP_001321134.1 | 3 - 4 | 764 | 81 | |
NM_101993.4 | NP_564139.1 | 3 - 4 | 838 | 81 | ||
GenBank mRNA | AY058081.1 | - | 3 - 4 | 695 | 81 | |
AY090308.1 | - | 2 - 3 | 563 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |