Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATGRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345737.1 | NP_001332639.1 | 19 - 20 | 2373 | 67 | |
NM_001345738.1 | NP_001332640.1 | 18 - 19 | 2419 | 67 | ||
NM_126000.2 | NP_569025.2 | 18 - 19 | 2078 | 67 | ||
NM_180946.3 | NP_851277.1 | 19 - 20 | 2408 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |