Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126014.4 | NP_201417.1 | 2 - 3 | 479 | 100 | |
NM_203272.2 | NP_975001.1 | 2 - 3 | 479 | 100 | ||
GenBank mRNA | AF218807.1 | - | 2 - 3 | 484 | 100 | |
AY035089.1 | - | 2 - 3 | 414 | 100 | ||
AY051036.1 | - | 2 - 3 | 367 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |