Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085337.1 | NP_001078806.1 | 4 - 5 | 762 | 84 | |
NM_001126036.2 | NP_001119508.1 | 4 - 5 | 762 | 84 | ||
NM_001345751.1 | NP_001331199.1 | 4 - 5 | 762 | 84 | ||
NM_001345752.1 | NP_001331200.1 | 4 - 5 | 792 | 84 | ||
NM_126019.4 | NP_201422.1 | 5 - 6 | 654 | 84 | ||
NM_180949.3 | NP_851280.1 | 4 - 5 | 762 | 84 | ||
GenBank mRNA | AK227052.1 | - | 4 - 5 | 587 | 84 | |
AK317109.1 | - | 4 - 5 | 581 | 84 | ||
AY139991.1 | - | 4 - 5 | 586 | 84 | ||
BT010380.1 | - | 4 - 5 | 572 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |