Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126039.4 | NP_569033.1 | 4 - 5 | 823 | 68 | |
GenBank mRNA | AY086393.1 | - | 4 - 5 | 454 | 68 | |
BT003998.1 | - | 4 - 5 | 490 | 68 | ||
BT005683.1 | - | 4 - 5 | 351 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |