Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMMA CA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085339.1 | NP_001078808.1 | 3 - 4 | 530 | 106 | |
NM_126049.4 | NP_569036.1 | 3 - 4 | 529 | 106 | ||
GenBank mRNA | AY045784.1 | - | 3 - 4 | 497 | 106 | |
AY079385.1 | - | 3 - 4 | 389 | 106 | ||
AY087379.1 | - | 3 - 4 | 524 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |