Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKKK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345806.1 | NP_001330515.1 | 5 - 6 | 1672 | 75 | |
NM_126084.3 | NP_569040.1 | 5 - 6 | 1672 | 75 | ||
GenBank mRNA | AF360242.1 | - | 5 - 6 | 1570 | 75 | |
AY040040.1 | - | 5 - 6 | 1405 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |