Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345811.1 | NP_001331725.1 | 24 - 25 | 3936 | 90 | |
NM_126110.3 | NP_201511.2 | 24 - 25 | 3918 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |