Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126111.4 | NP_201512.1 | 3 - 4 | 473 | 98 | |
GenBank mRNA | BT028946.1 | - | 3 - 4 | 380 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |