Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126128.4 | NP_201529.1 | 1 - 2 | 1781 | 66 | |
GenBank mRNA | AK227163.1 | - | 1 - 2 | 1678 | 66 | |
BT001094.1 | - | 1 - 2 | 1613 | 66 | ||
FJ708818.1 | - | 1 - 2 | 1613 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |