Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126132.4 | NP_201533.1 | 13 - 14 | 1734 | 66 | |
GenBank mRNA | AY057698.1 | - | 13 - 14 | 1734 | 66 | |
AY143932.1 | - | 13 - 14 | 1620 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |