Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126043.1 | NP_001119515.1 | 3 - 4 | 1956 | 101 | |
NM_001345830.1 | NP_001331087.1 | 4 - 5 | 1655 | 101 | ||
NM_126144.4 | NP_201545.1 | 4 - 5 | 2037 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |