Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QSOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335047.1 | NP_001324094.1 | 3 - 4 | 901 | 85 | |
NM_001335048.1 | NP_001324093.1 | 4 - 5 | 784 | 85 | ||
NM_126188.4 | NP_565258.1 | 4 - 5 | 636 | 85 | ||
GenBank mRNA | AY090364.1 | - | 4 - 5 | 610 | 85 | |
AY125539.1 | - | 4 - 5 | 488 | 85 | ||
BT000880.1 | - | 4 - 5 | 488 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |