Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QSOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335047.1 | NP_001324094.1 | 4 - 5 | 969 | 72 | |
NM_001335048.1 | NP_001324093.1 | 5 - 6 | 852 | 72 | ||
NM_126188.4 | NP_565258.1 | 5 - 6 | 704 | 72 | ||
GenBank mRNA | AY090364.1 | - | 5 - 6 | 678 | 72 | |
AY125539.1 | - | 5 - 6 | 556 | 72 | ||
BT000880.1 | - | 5 - 6 | 556 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |