Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335052.1 | NP_001325195.1 | 7 - 8 | 497 | 106 | |
NM_001335053.1 | NP_001325196.1 | 7 - 8 | 497 | 106 | ||
NM_126189.4 | NP_178237.3 | 7 - 8 | 497 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |