Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXGT-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126246.4 | NP_178294.1 | 2 - 3 | 480 | 174 | |
GenBank mRNA | AY059910.1 | - | 2 - 3 | 428 | 174 | |
BT008820.1 | - | 2 - 3 | 277 | 174 | ||
D63509.1 | - | 2 - 3 | 416 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |