Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126249.5 | NP_178297.2 | 3 - 4 | 668 | 70 | |
GenBank mRNA | AF492659.1 | - | 3 - 4 | 451 | 70 | |
BT026449.1 | - | 3 - 4 | 451 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |