Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMAP65-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035868.2 | NP_001030945.1 | 10 - 11 | 1454 | 71 | |
NM_126252.4 | NP_178300.3 | 10 - 11 | 2641 | 71 | ||
GenBank mRNA | AK317525.1 | - | 10 - 11 | 1752 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |