Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMAP65-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035868.2 | NP_001030945.1 | 7 - 8 | 1223 | 100 | |
NM_126252.4 | NP_178300.3 | 7 - 8 | 2410 | 100 | ||
GenBank mRNA | AK317525.1 | - | 7 - 8 | 1521 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |