Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMAP65-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035868.2 | NP_001030945.1 | 9 - 10 | 1374 | 94 | |
NM_126252.4 | NP_178300.3 | 9 - 10 | 2561 | 94 | ||
GenBank mRNA | AK317525.1 | - | 9 - 10 | 1672 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |