Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035872.3 | NP_001030949.2 | 2 - 3 | 568 | 89 | |
NM_126255.4 | NP_178303.2 | 2 - 3 | 1020 | 89 | ||
GenBank mRNA | AK226703.1 | - | 2 - 3 | 574 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |