Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335120.1 | NP_001324706.1 | 2 - 3 | 2365 | 124 | |
NM_126303.3 | NP_565285.1 | 2 - 3 | 2354 | 124 | ||
GenBank mRNA | AF264023.1 | - | 2 - 3 | 2231 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |