Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335131.1 | NP_001318183.1 | 1 - 2 | 987 | 66 | |
NM_001335132.1 | NP_001324185.1 | 1 - 2 | 987 | 66 | ||
NM_126326.5 | NP_565288.1 | 1 - 2 | 1027 | 66 | ||
NM_179597.1 | NP_849928.1 | 1 - 2 | 976 | 66 | ||
GenBank mRNA | AB038798.1 | - | 1 - 2 | 977 | 66 | |
AY045642.1 | - | 1 - 2 | 779 | 66 | ||
AY058232.1 | - | 1 - 2 | 571 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |