Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335144.1 | NP_001324495.1 | 5 - 6 | 1261 | 75 | |
NM_001335145.1 | NP_001324494.1 | 5 - 6 | 804 | 75 | ||
NM_126334.3 | NP_178382.2 | 5 - 6 | 788 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |