Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111077.4 | NP_186860.1 | 1 - 2 | 440 | 104 | |
GenBank mRNA | AY037210.1 | - | 1 - 2 | 327 | 104 | |
BT002657.1 | - | 1 - 2 | 272 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |