Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337384.1 | NP_001327088.1 | 3 - 4 | 2496 | 86 | |
NM_111084.4 | NP_566165.2 | 4 - 5 | 2566 | 86 | ||
GenBank mRNA | AY150501.1 | - | 4 - 5 | 1999 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |