Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337388.1 | NP_001327521.1 | 3 - 4 | 758 | 66 | |
NM_001337390.1 | NP_001327520.1 | 3 - 4 | 629 | 66 | ||
NM_111093.2 | NP_186875.2 | 3 - 4 | 597 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |